The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321701.1 | NP_001308630.1 | 2 - 3 | 154 | 169 |
NR_135762.1 | 2 - 3 | 154 | 169 | ||
XM_017027112.1 | XP_016882601.1 | 2 - 3 | 455 | 169 | |
GenBank mRNA | BG911482.1 | - | 2 - 3 | 141 | 169 |
CD389050.1 | - | 2 - 3 | 176 | 169 |