Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A25
Entrez Gene ID
387601
Gene Name
solute carrier family 22 member 25
Gene Aliases
HIMTP, UST6
Chromosome Location
Chr.11: 63162264 - 63243523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199352.3 NP_955384.3 1 - 2 402 146
XM_017017684.1 XP_016873173.1 4 - 5 1481 146
XM_017017685.1 XP_016873174.1 5 - 6 1609 146
XM_017017686.1 XP_016873175.1 4 - 5 1917 146
XM_017017687.1 XP_016873176.1 4 - 5 2022 146
XM_017017695.1 XP_016873184.1 4 - 5 2022 146
GenBank mRNA AK127610.1 - 1 - 2 277 146
AY437532.1 - 1 - 2 402 146
BC101315.1 - 1 - 2 397 146
BC101316.1 - 1 - 2 402 146

View Full Product Details