The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077594.1 | NP_001071062.1 | 9 - 10 | 1929 | 142 |
XM_011537323.2 | XP_011535625.1 | 10 - 11 | 2604 | 142 | |
XM_011537324.2 | XP_011535626.1 | 10 - 11 | 2588 | 142 | |
XM_011537325.2 | XP_011535627.1 | 11 - 12 | 2360 | 142 | |
XM_011537327.2 | XP_011535629.1 | 10 - 11 | 2059 | 142 | |
XM_011537328.2 | XP_011535630.1 | 11 - 12 | 2091 | 142 | |
XM_011537329.2 | XP_011535631.1 | 11 - 12 | 2499 | 142 | |
XM_011537330.2 | XP_011535632.1 | 12 - 13 | 2280 | 142 | |
XM_011537331.1 | XP_011535633.1 | 11 - 12 | 2075 | 142 | |
XM_011537332.2 | XP_011535634.1 | 10 - 11 | 2304 | 142 | |
XM_011537333.2 | XP_011535635.1 | 11 - 12 | 2336 | 142 | |
XM_011537334.1 | XP_011535636.1 | 9 - 10 | 2287 | 142 | |
GenBank mRNA | AK000671.1 | - | 6 - 7 | 1701 | 142 |
BC113655.1 | - | 9 - 10 | 1929 | 142 | |
BC117375.1 | - | 9 - 10 | 1929 | 142 |