The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077594.1 | NP_001071062.1 | 3 - 4 | 1236 | 66 |
XM_011537323.2 | XP_011535625.1 | 4 - 5 | 1911 | 66 | |
XM_011537324.2 | XP_011535626.1 | 4 - 5 | 1913 | 66 | |
XM_011537325.2 | XP_011535627.1 | 5 - 6 | 1667 | 66 | |
XM_011537327.2 | XP_011535629.1 | 4 - 5 | 1366 | 66 | |
XM_011537328.2 | XP_011535630.1 | 5 - 6 | 1398 | 66 | |
XM_011537329.2 | XP_011535631.1 | 5 - 6 | 1806 | 66 | |
XM_011537330.2 | XP_011535632.1 | 6 - 7 | 1587 | 66 | |
XM_011537331.1 | XP_011535633.1 | 5 - 6 | 1382 | 66 | |
XM_011537332.2 | XP_011535634.1 | 4 - 5 | 1611 | 66 | |
XM_011537333.2 | XP_011535635.1 | 5 - 6 | 1643 | 66 | |
XM_011537334.1 | XP_011535636.1 | 3 - 4 | 1594 | 66 | |
GenBank mRNA | AK000671.1 | - | 2 - 3 | 507 | 66 |
BC113655.1 | - | 3 - 4 | 1236 | 66 | |
BC117375.1 | - | 3 - 4 | 1236 | 66 |