Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A25
Entrez Gene ID
114789
Gene Name
solute carrier family 25 member 25
Gene Aliases
MCSC, PCSCL, SCAMC-2
Chromosome Location
Chr.9: 128068200 - 128109258 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006641.3 NP_001006642.1 4 - 5 707 151
NM_001006642.3 NP_001006643.1 4 - 5 952 151
NM_052901.4 NP_443133.2 4 - 5 570 151
NR_049766.2 4 - 5 613 151
XM_005251689.4 XP_005251746.1 4 - 5 966 151
GenBank mRNA AJ619963.1 - 4 - 5 871 151
AJ619989.1 - 4 - 5 495 151
AJ619990.1 - 4 - 5 590 151
AJ619991.1 - 4 - 5 552 151
AJ619992.1 - 4 - 5 538 151
AK290705.1 - 4 - 5 543 151
AK290991.1 - 4 - 5 707 151
AL559911.3 - 4 - 5 715 151
AY358515.1 - 4 - 5 545 151
BC089448.1 - 4 - 5 561 151
BC103930.1 - 4 - 5 512 151
BC103931.1 - 4 - 5 512 151
BC103932.1 - 4 - 5 512 151
BC103933.1 - 4 - 5 512 151

View Full Product Details