The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017004223.1 | XP_016859712.1 | 1 - 1 | 553 | 120 |
GenBank mRNA | BC127903.1 | - | 1 - 1 | 240 | 120 |