Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRAMD3
Entrez Gene ID
65983
Gene Name
GRAM domain containing 3
Gene Aliases
NS3TP2
Chromosome Location
Chr.5: 126360096 - 126494377 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146319.1 NP_001139791.1 3 - 4 600 99
NM_001146321.1 NP_001139793.1 4 - 5 792 99
NM_001146322.1 NP_001139794.1 3 - 4 411 99
NM_023927.2 NP_076416.2 3 - 4 749 99
XM_005272057.3 XP_005272114.1 3 - 4 487 99
XM_005272058.3 XP_005272115.1 3 - 4 487 99
XM_005272059.4 XP_005272116.1 4 - 5 877 99
XM_005272060.3 XP_005272117.1 3 - 4 574 99
XM_005272061.3 XP_005272118.1 3 - 4 569 99
XM_005272062.3 XP_005272119.1 3 - 4 535 99
XM_005272063.3 XP_005272120.1 3 - 4 684 99
XM_005272064.3 XP_005272121.1 4 - 5 594 99
XM_011543591.2 XP_011541893.1 4 - 5 1465 99
XM_011543592.2 XP_011541894.1 4 - 5 1461 99
XM_011543593.2 XP_011541895.1 3 - 4 574 99
XM_011543594.2 XP_011541896.1 3 - 4 346 99
XM_017009781.1 XP_016865270.1 4 - 5 352 99
XM_017009782.1 XP_016865271.1 4 - 5 594 99
GenBank mRNA AK024966.1 - 3 - 4 517 99
AK225829.1 - 3 - 4 517 99
AK296267.1 - 4 - 5 497 99
AK298009.1 - 3 - 4 334 99
AK298349.1 - 3 - 4 422 99
AK300154.1 - 3 - 4 545 99
AK303857.1 - 3 - 4 351 99
AK316288.1 - 3 - 4 564 99
AY116970.1 - 3 - 4 319 99
BC008590.1 - 3 - 4 482 99

View Full Product Details