Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GRAMD3
- Entrez Gene ID
-
65983
- Gene Name
- GRAM domain containing 3
- Gene Aliases
-
NS3TP2
- Chromosome Location
-
Chr.5: 126360096 - 126494377 on Build GRCh38
View Full Product Details