Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSTA3
Entrez Gene ID
7264
Gene Name
tissue specific transplantation antigen P35B
Gene Aliases
FX, P35B, SDR4E1
Chromosome Location
Chr.8: 143612618 - 143618043 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317783.1 NP_001304712.1 8 - 9 792 129
NM_003313.3 NP_003304.1 8 - 9 829 129
XM_005251051.3 XP_005251108.2 8 - 9 764 129
XM_011517269.1 XP_011515571.1 8 - 9 856 129
GenBank mRNA AK096752.1 - 6 - 7 1691 129
AK313560.1 - 8 - 9 771 129
BC001941.1 - 8 - 9 765 129
BC093061.1 - 8 - 9 806 129
U58766.1 - 7 - 8 803 129

View Full Product Details