The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191013.1 | NP_001177942.1 | 4 - 5 | 993 | 114 |
NM_001191015.1 | NP_001177944.1 | 2 - 3 | 428 | 114 | |
GenBank mRNA | AK296266.1 | - | 3 - 4 | 380 | 114 |
BM708208.1 | - | 2 - 3 | 332 | 114 | |
DC341647.1 | - | 3 - 4 | 380 | 114 |