The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017020213.1 | XP_016875702.1 | 5 - 6 | 603 | 111 |
XM_017020214.1 | XP_016875703.1 | 5 - 6 | 603 | 111 |