The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320178.1 | NP_001307107.1 | 3 - 4 | 509 | 111 |
NM_001320179.1 | NP_001307108.1 | 3 - 4 | 806 | 111 | |
NM_198494.2 | NP_940896.2 | 3 - 4 | 509 | 111 | |
XM_005270809.4 | XP_005270866.1 | 3 - 4 | 1236 | 111 | |
XM_006710605.3 | XP_006710668.1 | 3 - 4 | 1216 | 111 | |
XM_006710606.3 | XP_006710669.1 | 3 - 4 | 1236 | 111 | |
GenBank mRNA | AK122618.1 | - | 3 - 4 | 509 | 111 |
BC041873.1 | - | 3 - 4 | 1222 | 111 | |
BC144174.1 | - | 2 - 3 | 255 | 111 | |
CV374692.1 | - | 1 - 2 | 75 | 111 |