Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK022419.1 - 1 1842 Chr. 1 - 63547082 - 63549558 [+] on Build GRCh38 94


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream EF-hand calcium binding domain 7 EFCAB7 Sense Chr. - 63523301 - 63585400 on Build GRCh38
Downstream phosphoglucomutase 1 PGM1 CDG1T, GSD14 43.7kb Sense Chr. - 63593276 - 63660245 on Build GRCh38

View Full Product Details