Product Details

Assay Reference Genome
Location

Chr.5:148992453 on build GRCh38
Cytoband
5q32
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
79628
Gene Symbol
SH3TC2
Gene Name
SH3 domain and tetratricopeptide repeats 2
Gene Aliases
CMT4C, MNMN
Location
Chr.5:148982150-149063174 on build GRCh38
Assay Gene Location
Within Exon 17
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
SH3TC2 NM_024577.3 17 16277 NP_078853.2

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv830517 Chr5:148953659 - 149171578 on Build GRCh38 Gain LOC255187 MIR584 SH3TC2 ABLIM3

View Full Product Details