Product Details
- Assay Reference Genome
Location
Chr.2:218278533 on build GRCh38
- Cytoband
- 2q35
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 25953
- Gene Symbol
- PNKD
- Gene Name
- paroxysmal nonkinesigenic dyskinesia
- Gene Aliases
- BRP17, DYT8, FKSG19, FPD1, KIPP1184, MR-1, MR1, PDC, PKND1, TAHCCP2
- Location
- Chr.2:218270392-218346793 on build GRCh38
- Assay Gene Location
- Within Intron 3
- Entrez Gene ID
- 64114
- Gene Symbol
- TMBIM1
- Gene Name
- transmembrane BAX inhibitor motif containing 1
- Gene Aliases
- LFG3, MST100, MSTP100, PP1201, RECS1
- Location
- Chr.2:218274192-218292577 on build GRCh38
- Assay Gene Location
- Overlaps Intron 10 - Intron 11
View Full Product Details