The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000686.4 | NP_000677.2 | 3 - 3 | 2443 | 165 |
XM_011537533.1 | XP_011535835.1 | 2 - 2 | 2341 | 165 |