Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HCP5
Entrez Gene ID
10866
Gene Name
HLA complex P5 (non-protein coding)
Gene Aliases
6S2650E, D6S2650E, P5-1
Chromosome Location
Chr.6: 31463180 - 31465809 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_040662.1 2 - 2 2357 113
GenBank mRNA AK093953.1 - 4 - 4 2242 113
BX640979.1 - 2 - 2 2355 113

View Full Product Details