Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FLRT3
Entrez Gene ID
23767
Gene Name
fibronectin leucine rich transmembrane protein 3
Gene Aliases
HH21
Chromosome Location
Chr.20: 14322985 - 14337847 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013281.3 NP_037413.1 2 - 2 3509 140
NM_198391.2 NP_938205.1 3 - 3 3703 140
XM_005260682.4 XP_005260739.1 3 - 3 4221 140
XM_011529204.2 XP_011527506.1 4 - 4 4358 140
XM_011529205.2 XP_011527507.1 4 - 4 4020 140
GenBank mRNA AB040902.1 - 2 - 2 3397 140
AK027670.1 - 1 - 1 2617 139
AK074909.1 - 2 - 2 3377 139

View Full Product Details