Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRG3
Entrez Gene ID
10718
Gene Name
neuregulin 3
Gene Aliases
HRG3, pro-NRG3
Chromosome Location
Chr.10: 81875298 - 82989979 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010848.3 NP_001010848.2 7 - 8 1441 74
NM_001165972.1 NP_001159444.1 7 - 8 1438 74
NM_001165973.1 NP_001159445.1 8 - 9 1025 74
XM_005269444.4 XP_005269501.1 7 - 8 1442 74
XM_017015574.1 XP_016871063.1 8 - 9 1522 74
XM_017015576.1 XP_016871065.1 8 - 9 1519 74
XM_017015578.1 XP_016871067.1 8 - 9 1522 74
XM_017015580.1 XP_016871069.1 8 - 9 1519 74
XM_017015583.1 XP_016871072.1 8 - 9 794 74
GenBank mRNA AK098823.1 - 6 - 7 585 74
AK122886.1 - 9 - 10 1280 74
AK296563.1 - 7 - 8 1263 74
BC136811.1 - 7 - 8 1438 74
BC143665.1 - 5 - 6 475 74
DQ001411.1 - 7 - 8 1414 74
DQ857894.1 - 8 - 9 1027 74
HM068873.1 - 7 - 8 1414 74
HM068874.1 - 5 - 6 1184 74
HM068875.1 - 8 - 9 751 74
HM068876.1 - 7 - 8 651 74
HM068878.1 - 9 - 10 782 74
HM068879.1 - 7 - 8 651 74
HM068880.1 - 7 - 8 726 74
HM068881.1 - 9 - 10 857 74
HM068882.1 - 8 - 9 826 74
HM068883.1 - 8 - 9 826 74
HM068884.1 - 9 - 10 904 74
HM068885.1 - 5 - 6 497 74

View Full Product Details