Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TCEAL6
Entrez Gene ID
158931
Gene Name
transcription elongation factor A like 6
Gene Aliases
Tceal3, WEX2
Chromosome Location
Chr.X: 102139961 - 102142416 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006938.2 NP_001006939.2 3 - 3 770 95
GenBank mRNA BC071675.1 - 3 - 3 754 95

View Full Product Details