Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRTAP10-2
Entrez Gene ID
386679
Gene Name
keratin associated protein 10-2
Gene Aliases
KAP10.2, KAP18-2, KAP18.2, KRTAP10.2, KRTAP18-2, KRTAP18.2
Chromosome Location
Chr.21: 44550357 - 44551505 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198693.3 NP_941966.1 1 - 1 727 115
GenBank mRNA AB076348.1 - 1 - 1 727 115

View Full Product Details