The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005170.2 | NP_001005170.1 | 1 - 1 | 165 | 152 |
GenBank mRNA | BC136921.1 | - | 1 - 1 | 165 | 152 |
BC151144.1 | - | 1 - 1 | 165 | 152 |