The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002261.2 | NP_002252.2 | 6 - 6 | 656 | 146 |
NM_007333.2 | NP_031359.2 | 6 - 6 | 656 | 146 | |
GenBank mRNA | AF078550.1 | - | 6 - 6 | 611 | 146 |
AF350016.1 | - | 6 - 6 | 611 | 146 | |
AF350017.1 | - | 6 - 6 | 611 | 146 | |
AF461157.1 | - | 6 - 6 | 627 | 146 | |
L14542.1 | - | 6 - 6 | 656 | 146 |