The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085386.1 | NP_001078855.1 | 1 - 2 | 336 | 101 |
GenBank mRNA | AY258907.1 | - | 1 - 2 | 336 | 101 |