Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
NPM1
Entrez Gene ID
4869
Gene Name
nucleophosmin
Gene Aliases
B23, NPM
Chromosome Location
Chr.5: 171387648 - 171410900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY740635.1 - 11 - 11 939 147

View Full Product Details