Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAP1
Entrez Gene ID
10487
Gene Name
adenylate cyclase associated protein 1
Gene Aliases
CAP, CAP1-PEN
Chromosome Location
Chr.1: 40040070 - 40072649 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105530.1 NP_001099000.1 7 - 8 842 90
NM_006367.3 NP_006358.1 7 - 8 860 90
XM_005270368.1 XP_005270425.1 7 - 8 700 90
XM_011540509.1 XP_011538811.1 8 - 9 1253 90
XM_011540510.1 XP_011538812.1 8 - 9 1250 90
XM_011540511.1 XP_011538813.1 7 - 8 684 90
XM_011540512.1 XP_011538814.1 7 - 8 803 90
XM_011540513.2 XP_011538815.1 7 - 8 695 90
XM_011540514.1 XP_011538816.1 8 - 9 810 90
XM_011540515.1 XP_011538817.1 8 - 9 1250 90
XM_017000067.1 XP_016855556.1 7 - 8 768 90
XM_017000068.1 XP_016855557.1 8 - 9 812 90
XM_017000069.1 XP_016855558.1 8 - 9 1023 90
XM_017000070.1 XP_016855559.1 7 - 8 1042 90
XM_017000071.1 XP_016855560.1 7 - 8 1000 90
XM_017000072.1 XP_016855561.1 7 - 8 936 90
XM_017000073.1 XP_016855562.1 7 - 8 757 90
GenBank mRNA AK222513.1 - 7 - 8 701 90
AK295396.1 - 8 - 9 622 90
AK297829.1 - 7 - 8 569 90
AK298094.1 - 7 - 8 540 90
AK298110.1 - 7 - 8 644 90
AK315731.1 - 7 - 8 747 90
BC013963.2 - 7 - 8 677 90
BC095440.1 - 7 - 8 682 90
BT007152.1 - 6 - 7 631 90
CR457409.1 - 6 - 7 631 90
L12168.1 - 6 - 7 670 90
M98474.1 - 7 - 8 701 90

View Full Product Details