The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136035.2 | NP_001129507.1 | 6 - 7 | 891 | 82 |
NM_001142554.1 | NP_001136026.1 | 5 - 6 | 1012 | 82 | |
NM_017722.3 | NP_060192.1 | 5 - 6 | 1012 | 82 | |
XM_006722793.2 | XP_006722856.1 | 5 - 6 | 1134 | 82 | |
XM_011528124.1 | XP_011526426.1 | 6 - 7 | 810 | 82 | |
XM_011528125.1 | XP_011526427.1 | 7 - 8 | 1343 | 82 | |
XM_017026944.1 | XP_016882433.1 | 7 - 8 | 1334 | 82 | |
XM_017026945.1 | XP_016882434.1 | 7 - 8 | 1334 | 82 | |
XM_017026946.1 | XP_016882435.1 | 5 - 6 | 1134 | 82 | |
GenBank mRNA | AF196479.1 | - | 5 - 6 | 762 | 82 |
AK000251.1 | - | 6 - 7 | 850 | 82 | |
AK292377.1 | - | 6 - 7 | 891 | 82 | |
AK295245.1 | - | 6 - 7 | 791 | 82 | |
AK304636.1 | - | 6 - 7 | 831 | 82 | |
BC002492.2 | - | 5 - 6 | 842 | 82 | |
BC018302.1 | - | 5 - 6 | 1007 | 82 | |
BC040126.1 | - | 5 - 6 | 881 | 82 |