The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136035.2 | NP_001129507.1 | 7 - 8 | 1002 | 87 |
NM_001142554.1 | NP_001136026.1 | 6 - 7 | 1123 | 87 | |
NM_017722.3 | NP_060192.1 | 6 - 7 | 1123 | 87 | |
XM_006722793.2 | XP_006722856.1 | 6 - 7 | 1245 | 87 | |
XM_011528124.1 | XP_011526426.1 | 7 - 8 | 921 | 87 | |
XM_011528125.1 | XP_011526427.1 | 8 - 9 | 1454 | 87 | |
XM_017026944.1 | XP_016882433.1 | 8 - 9 | 1445 | 87 | |
XM_017026945.1 | XP_016882434.1 | 8 - 9 | 1445 | 87 | |
XM_017026946.1 | XP_016882435.1 | 6 - 7 | 1245 | 87 | |
XM_017026947.1 | XP_016882436.1 | 8 - 9 | 1431 | 87 | |
GenBank mRNA | AF196479.1 | - | 6 - 7 | 873 | 87 |
AK000251.1 | - | 7 - 8 | 961 | 87 | |
AK292377.1 | - | 7 - 8 | 1002 | 87 | |
AK295245.1 | - | 7 - 8 | 902 | 87 | |
AK304636.1 | - | 7 - 8 | 942 | 87 | |
BC002492.2 | - | 6 - 7 | 953 | 87 | |
BC018302.1 | - | 6 - 7 | 1118 | 87 | |
BC040126.1 | - | 6 - 7 | 992 | 87 |