Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HNRNPA3P1
Entrez Gene ID
10151
Gene Name
heterogeneous nuclear ribonucleoprotein A3 pseudogene 1
Gene Aliases
D10S102, FBRNP, HNRPA3, HNRPA3P1
Chromosome Location
Chr.10: 43787412 - 43790417 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_002726.2 1 - 1 2455 99


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC102724264 LOC102724264 -8.6kb Sense Chr. - 43777562 - 43778902 on Build GRCh38
Downstream long intergenic non-protein coding RNA 619 LINC00619 C10orf136, bA168P8.1 54.9kb Anti-sense Chr. - 43845306 - 43850622 on Build GRCh38

View Full Product Details