The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011510158.2 | XP_011508460.1 | 2 - 2 | 5257 | 59 |
GenBank mRNA | AL050068.1 | - | 1 - 1 | 154 | 59 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | ADP ribosylation factor like GTPase 8A | ARL8A | ARL10B, GIE2 | Anti-sense | Chr. - 202133404 - 202144743 on Build GRCh38 | |
Downstream | protein tyrosine phosphatase, non-receptor type 7 | PTPN7 | BPTP-4, HEPTP, LC-PTP, LPTP, PTPNI | 13.4kb | Anti-sense | Chr. - 202147012 - 202161588 on Build GRCh38 |