Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BC127210.1 |
- |
1 |
151 |
|
Chr. 14 - 68556514 - 68556855 [+] on Build GRCh38 |
83 |
GenBankmRNA |
CB853664.1 |
- |
1 |
589 |
|
Chr. 14 - 68556076 - 68556812 [+] on Build GRCh38 |
83 |
GenBankmRNA |
CF891492.1 |
- |
1 |
589 |
|
Chr. 14 - 68556076 - 68556865 [+] on Build GRCh38 |
83 |
GenBankmRNA |
CB853447.1 |
- |
1 |
589 |
|
Chr. 14 - 68556076 - 68556873 [+] on Build GRCh38 |
83 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
zinc finger FYVE-type containing 26 |
ZFYVE26 |
FYVE-CENT, SPG15 |
-739.9kb
|
Anti-sense |
Chr. - 67728892 - 67817293 on Build GRCh38 |
Downstream |
uncharacterized LOC100996664 |
LOC100996664 |
|
48.4kb
|
Anti-sense |
Chr. - 68625527 - 68628787 on Build GRCh38 |
View Full Product Details