Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PMP22
- Entrez Gene ID
-
5376
- Gene Name
- peripheral myelin protein 22
- Gene Aliases
-
CMT1A, CMT1E, DSS, GAS-3, GAS3, HMSNIA, HNPP, Sp110
- Chromosome Location
-
Chr.17: 15229777 - 15265357 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AL833462.1 |
- |
1 - 1 |
3850 |
68 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA583529.1 |
- |
1 |
262 |
|
Chr. 17 - 15240413 - 15240802 [-] on Build GRCh38 |
68 |
GenBankmRNA |
BX470817.1 |
- |
1 |
|
|
Chr. 17 - 15240407 - 15240891 [+] on Build GRCh38 |
68 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
CMT1A duplicated region transcript 8 |
CDRT8 |
|
-134.2kb
|
Sense |
Chr. - 15104985 - 15106187 on Build GRCh38 |
Downstream |
microRNA 4731 |
MIR4731 |
|
7.2kb
|
Sense |
Chr. - 15251627 - 15251696 on Build GRCh38 |
View Full Product Details