Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FXR1
Entrez Gene ID
8087
Gene Name
FMR1 autosomal homolog 1
Gene Aliases
FXR1P
Chromosome Location
Chr.3: 180912446 - 180982753 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013438.2 NP_001013456.1 16 - 16 8161 98
NM_001013439.2 NP_001013457.1 18 - 18 8597 98
NM_005087.3 NP_005078.2 17 - 17 8253 98
GenBank mRNA AF075028.1 - 1 - 1 248 98

View Full Product Details