Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL15
Entrez Gene ID
6138
Gene Name
ribosomal protein L15
Gene Aliases
DBA12, EC45, L15, RPL10, RPLY10, RPYL10
Chromosome Location
Chr.3: 23916804 - 23923696 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253379.1 NP_001240308.1 3 - 4 707 93
NM_001253380.1 NP_001240309.1 2 - 3 469 93
NM_001253382.1 NP_001240311.1 3 - 4 458 93
NM_001253383.1 NP_001240312.1 3 - 4 437 93
NM_002948.3 NP_002939.2 3 - 4 694 93
GenBank mRNA AF283772.2 - 3 - 4 354 93
AK091751.1 - 1 - 2 1434 93
AK098795.1 - 3 - 4 344 93
AK129815.1 - 3 - 4 354 93
AK290694.1 - 3 - 4 350 93
AK292000.1 - 3 - 4 350 93
AY347528.1 - 2 - 3 419 93
BC014837.1 - 3 - 4 357 93
BC068198.1 - 1 - 1 353 93
BC070328.1 - 3 - 4 350 93
BC071672.1 - 1 - 1 339 93
BE893727.1 - 3 - 4 429 93
BT019672.1 - 2 - 3 314 93
BU178524.1 - 3 - 4 458 93
DA037869.1 - 3 - 4 375 93
DA205054.1 - 3 - 4 420 93
L25899.1 - 2 - 3 330 93

View Full Product Details