Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- H2AFY
- Entrez Gene ID
-
9555
- Gene Name
- H2A histone family member Y
- Gene Aliases
-
H2A.y, H2A/y, H2AF12M, MACROH2A1.1, mH2A1, macroH2A1.2
- Chromosome Location
-
Chr.5: 135334381 - 135399887 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CX167042.1 |
- |
1 |
|
|
Chr. 5 - 135389107 - 135399533 [-] on Build GRCh38 |
106 |
GenBankmRNA |
DA148558.1 |
- |
1 |
|
|
Chr. 5 - 135399165 - 135399622 [-] on Build GRCh38 |
106 |
GenBankmRNA |
DA274581.1 |
- |
1 |
157 |
|
Chr. 5 - 135399286 - 135399842 [+] on Build GRCh38 |
106 |
GenBankmRNA |
DA920818.1 |
- |
1 |
148 |
|
Chr. 5 - 135399280 - 135399836 [+] on Build GRCh38 |
106 |
GenBankmRNA |
BI560542.1 |
- |
1 |
82 |
|
Chr. 5 - 135399347 - 135400052 [+] on Build GRCh38 |
106 |
GenBankmRNA |
BQ948280.1 |
- |
|
134 |
|
Chr. - - [-] on Build |
106 |
GenBankmRNA |
AK092789.1 |
- |
1 |
148 |
|
Chr. 5 - 135399280 - 135401292 [+] on Build GRCh38 |
106 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
chromosome 5 open reading frame 66 |
C5orf66 |
|
-54.6kb
|
Sense |
Chr. - 135033280 - 135344680 on Build GRCh38 |
Downstream |
dendritic cell associated nuclear protein |
DCANP1 |
C5orf20, DCNP1 |
42.9kb
|
Anti-sense |
Chr. - 135444214 - 135447348 on Build GRCh38 |
View Full Product Details