Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ002451.1 - 1 Chr. 17 - 20303154 - 20303755 [-] on Build GRCh38 72
GenBankmRNA AK094434.1 - 1 1151 Chr. 17 - 20302368 - 20303952 [+] on Build GRCh38 72


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream sperm antigen with calponin homology and coiled-coil domains 1 SPECC1 CYTSB, HCMOGT-1, HCMOGT1, NSP Sense Chr. - 20009301 - 20319026 on Build GRCh38
Downstream coiled-coil domain containing 144C, pseudogene CCDC144CP CCDC144C 17.2kb Sense Chr. - 20321174 - 20402191 on Build GRCh38

View Full Product Details