Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NAF1
- Entrez Gene ID
-
92345
- Gene Name
- nuclear assembly factor 1 ribonucleoprotein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 163109133 - 163170388 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK126037.1 |
- |
1 - 1 |
2453 |
79 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BX103045.1 |
- |
2 |
306 |
|
Chr. 4 - 163110072 - 163111752 [-] on Build GRCh38 |
79 |
GenBankmRNA |
AA609049.1 |
- |
1 |
|
|
Chr. 4 - 163110072 - 163111720 [+] on Build GRCh38 |
79 |
GenBankmRNA |
AI652820.1 |
- |
1 |
|
|
Chr. 4 - 163110073 - 163140337 [+] on Build GRCh38 |
79 |
GenBankmRNA |
CN313742.1 |
- |
1 |
104 |
|
Chr. 4 - 163109648 - 163110281 [-] on Build GRCh38 |
79 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 4454 |
MIR4454 |
|
-15.5kb
|
Sense |
Chr. - 163093574 - 163093628 on Build GRCh38 |
Downstream |
neuropeptide Y receptor Y1 |
NPY1R |
NPY1-R, NPYR |
211.3kb
|
Sense |
Chr. - 163323962 - 163344847 on Build GRCh38 |
View Full Product Details