Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LOC101928514
- Entrez Gene ID
-
101928514
- Gene Name
- uncharacterized LOC101928514
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 76799044 - 76807102 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110837.1 |
|
2 - 2 |
1048 |
95 |
GenBank mRNA |
AK055500.1 |
- |
2 - 2 |
1048 |
95 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
major facilitator superfamily domain containing 11 |
MFSD11 |
ET |
|
Sense |
Chr. - 76736565 - 76803805 on Build GRCh38 |
Downstream |
mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, isozyme B |
MGAT5B |
GnT-IX, GnT-VB |
44.6kb
|
Sense |
Chr. - 76868487 - 76950389 on Build GRCh38 |
View Full Product Details