The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004580.4 | NP_004571.2 | 6 - 6 | 3230 | 105 |
NM_183234.2 | NP_899057.1 | 7 - 7 | 3211 | 105 | |
NM_183235.2 | NP_899058.1 | 7 - 7 | 3220 | 105 | |
NM_183236.2 | NP_899059.1 | 7 - 7 | 3171 | 105 | |
XM_005254576.4 | XP_005254633.1 | 6 - 6 | 4473 | 105 | |
XM_011521852.1 | XP_011520154.1 | 9 - 9 | 3434 | 105 | |
XM_011521854.1 | XP_011520156.1 | 8 - 8 | 3307 | 105 | |
XM_011521855.2 | XP_011520157.1 | 8 - 8 | 4696 | 105 | |
XM_011521856.2 | XP_011520158.1 | 6 - 6 | 3103 | 105 | |
GenBank mRNA | BC078139.1 | - | - | 2053 | 105 |
CA437806.1 | - | 1 - 1 | 105 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | ribosomal L24 domain containing 1 | RSL24D1 | C15orf15, HRP-L30-iso, L30, RLP24, RPL24, RPL24L, TVAS3 | -5.9kb | Sense | Chr. - 55181314 - 55197033 on Build GRCh38 |
Downstream | phosphatidylinositol glycan anchor biosynthesis class B | PIGB | GPI-MT-III, PIG-B | 113.2kb | Anti-sense | Chr. - 55318935 - 55355648 on Build GRCh38 |