Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CARF
- Entrez Gene ID
-
79800
- Gene Name
- calcium responsive transcription factor
- Gene Aliases
-
ALS2CR8, NYD-SP24
- Chromosome Location
-
Chr.2: 202912218 - 202987856 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK096293.1 |
- |
|
945 |
|
Chr. - - [-] on Build |
91 |
GenBankmRNA |
BM979415.1 |
- |
1 |
109 |
|
Chr. 2 - 202987360 - 202987725 [+] on Build GRCh38 |
91 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
WD repeat domain 12 |
WDR12 |
YTM1 |
-71.3kb
|
Anti-sense |
Chr. - 202876958 - 202912226 on Build GRCh38 |
Downstream |
neurobeachin like 1 |
NBEAL1 |
A530083I02Rik, ALS2CR16, ALS2CR17 |
26.8kb
|
Sense |
Chr. - 203014624 - 203225194 on Build GRCh38 |
View Full Product Details