Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AL110159.1 |
- |
1 |
552 |
|
Chr. 6 - 24700910 - 24701793 [-] on Build GRCh38 |
114 |
GenBankmRNA |
AI142111.1 |
- |
1 |
|
|
Chr. 6 - 24701039 - 24701600 [+] on Build GRCh38 |
114 |
GenBankmRNA |
AA669822.1 |
- |
1 |
|
|
Chr. 6 - 24700904 - 24701322 [+] on Build GRCh38 |
114 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
acyl-CoA thioesterase 13 |
ACOT13 |
HT012, PNAS-27, THEM2 |
|
Anti-sense |
Chr. - 24667035 - 24705069 on Build GRCh38 |
Downstream |
chromosome 6 open reading frame 62 |
C6orf62 |
Nbla00237, XTP12, dJ30M3.2 |
3.1kb
|
Sense |
Chr. - 24704758 - 24720836 on Build GRCh38 |
View Full Product Details