The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291485.1 | NP_001278414.1 | 3 - 4 | 817 | 67 |
NM_006890.4 | NP_008821.2 | 3 - 4 | 817 | 67 | |
GenBank mRNA | AK292213.1 | - | 3 - 4 | 810 | 67 |
AK296634.1 | - | 4 - 5 | 747 | 67 | |
AK313108.1 | - | 3 - 4 | 810 | 67 | |
BC121132.2 | - | 3 - 4 | 711 | 67 | |
L31792.1 | - | 3 - 4 | 707 | 67 | |
X98311.1 | - | 3 - 4 | 707 | 67 |