The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171581.1 | NP_001165052.1 | 3 - 4 | 753 | 88 |
NM_004867.4 | NP_004858.1 | 4 - 5 | 885 | 88 | |
GenBank mRNA | AF038953.1 | - | 4 - 5 | 687 | 88 |
AK223327.1 | - | 4 - 5 | 664 | 88 | |
AK303238.1 | - | 3 - 4 | 535 | 88 | |
AK313154.1 | - | 4 - 5 | 843 | 88 | |
AY359051.1 | - | 4 - 5 | 626 | 88 | |
BC040437.1 | - | 4 - 5 | 640 | 88 | |
BM561934.1 | - | 1 - 2 | 128 | 88 | |
CR456875.1 | - | 4 - 5 | 548 | 88 |