Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OR8S1
Entrez Gene ID
341568
Gene Name
olfactory receptor family 8 subfamily S member 1
Gene Aliases
-
Chromosome Location
Chr.12: 48525632 - 48528103 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005203.2 NP_001005203.2 1 - 2 884 85

View Full Product Details