Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CXorf40A
Entrez Gene ID
91966
Gene Name
chromosome X open reading frame 40A
Gene Aliases
CXorf40, EOLA1, FLJ16423
Chromosome Location
Chr.X: 149540630 - 149555338 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171907.2 NP_001165378.1 1 - 2 366 75
NM_001324276.1 NP_001311205.1 1 - 2 366 75
NM_178124.5 NP_835225.2 1 - 2 366 75
GenBank mRNA AK291899.1 - 1 - 2 292 75
AK294153.1 - 1 - 2 342 75
BC065539.1 - 1 - 2 291 75
BC104930.1 - 1 - 2 215 75
BC104932.1 - 1 - 2 215 75
BC143660.1 - 1 - 2 215 75
BC143661.1 - 1 - 2 215 75
BE265359.1 - 1 - 2 303 75
CB156531.1 - 1 - 2 366 75

View Full Product Details