Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DDN
Entrez Gene ID
23109
Gene Name
dendrin
Gene Aliases
-
Chromosome Location
Chr.12: 48995149 - 49001772 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015086.1 NP_055901.2 1 - 2 229 94
XM_011538055.2 XP_011536357.1 2 - 3 834 94
GenBank mRNA AB018292.1 - 1 - 2 113 94
BC136398.1 - 1 - 2 139 94
BC136399.1 - 1 - 2 139 94

View Full Product Details