The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145641.1 | NP_001139113.1 | 1 - 1 | 1946 | 95 |
GenBank mRNA | AK093056.1 | - | 1 - 1 | 1940 | 95 |
BC034980.1 | - | 2 - 2 | 1591 | 95 | |
BM679887.1 | - | 1 - 1 | 95 |