The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016038.2 | NP_057122.2 | 2 - 3 | 440 | 101 |
GenBank mRNA | AK001779.1 | - | 2 - 3 | 440 | 101 |
AK289609.1 | - | 2 - 3 | 445 | 101 | |
AY169963.1 | - | 2 - 3 | 440 | 101 | |
BC065700.1 | - | 2 - 3 | 410 | 101 | |
KU178629.1 | - | 2 - 3 | 256 | 101 |