The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321170.1 | NP_001308099.1 | 5 - 6 | 372 | 85 |
NM_138501.5 | NP_612510.1 | 4 - 5 | 298 | 85 | |
NR_038103.1 | 4 - 5 | 330 | 85 | ||
NR_038104.1 | 4 - 5 | 363 | 85 | ||
XM_006722945.1 | XP_006723008.1 | 5 - 6 | 416 | 85 | |
XM_011528442.2 | XP_011526744.1 | 4 - 5 | 246 | 85 | |
GenBank mRNA | AF038958.1 | - | 4 - 5 | 244 | 85 |
AF222742.1 | - | 4 - 5 | 219 | 85 | |
AK001416.1 | - | 4 - 5 | 262 | 85 | |
AK094087.1 | - | 5 - 6 | 339 | 85 | |
AK299134.1 | - | 5 - 6 | 336 | 85 | |
AK315412.1 | - | 4 - 5 | 262 | 85 | |
BC000174.2 | - | 3 - 4 | 158 | 85 | |
BC002425.1 | - | 4 - 5 | 230 | 85 | |
BC005952.1 | - | 4 - 5 | 241 | 85 | |
BC007801.2 | - | 4 - 5 | 248 | 85 | |
BC013881.2 | - | 4 - 5 | 246 | 85 | |
BG705196.1 | - | 4 - 5 | 301 | 85 | |
BT007179.1 | - | 3 - 4 | 168 | 85 | |
CR456892.1 | - | 3 - 4 | 168 | 85 |