Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TCP10L2
- Entrez Gene ID
-
401285
- Gene Name
- t-complex 10-like 2
- Gene Aliases
-
bA517H2.3
- Chromosome Location
-
Chr.6: 167170593 - 167196777 on Build GRCh38
- Gene Symbol
- TCP10
- Entrez Gene ID
-
6953
- Gene Name
- t-complex 10
- Gene Aliases
-
TCP10A
- Chromosome Location
-
Chr.6: 167357356 - 167384552 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC063451.1 |
- |
3 - 4 |
449 |
89 |
U03399.1 |
- |
3 - 4 |
439 |
89 |
View Full Product Details